Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052897.3 | NP_443129.3 | 9 - 10 | 2625 | 134 |
GenBank mRNA | AB067474.1 | - | 6 - 7 | 2294 | 134 |
AK056399.1 | - | 9 - 10 | 2659 | 134 | |
AL834230.1 | - | 3 - 4 | 904 | 134 | |
BC065530.1 | - | 9 - 10 | 2625 | 134 | |
CR627478.1 | - | 2 - 3 | 206 | 134 |