Assay Design



Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 11 - 12 1160 67
XM_005268725.3 XP_005268782.1 13 - 14 1510 67
XM_011538031.1 XP_011536333.1 13 - 14 1296 67
XM_011538033.1 XP_011536335.1 14 - 15 1350 67
XM_011538035.1 XP_011536337.1 13 - 14 1254 67
XM_011538036.1 XP_011536338.1 14 - 15 1350 67
XM_011538037.1 XP_011536339.1 12 - 13 1230 67
XM_011538038.1 XP_011536340.1 12 - 13 1200 67
XM_011538039.1 XP_011536341.1 13 - 14 1296 67
XM_011538040.1 XP_011536342.1 14 - 15 1350 67
XM_011538042.2 XP_011536344.1 15 - 16 1446 67
XM_017018998.1 XP_016874487.1 12 - 13 1272 67
XM_017018999.1 XP_016874488.1 15 - 16 1487 67
XM_017019000.1 XP_016874489.1 13 - 14 1254 67
XM_017019001.1 XP_016874490.1 14 - 15 1560 67
XM_017019002.1 XP_016874491.1 13 - 14 1326 67
XM_017019003.1 XP_016874492.1 12 - 13 1200 67
XM_017019004.1 XP_016874493.1 12 - 13 1194 67
XM_017019005.1 XP_016874494.1 14 - 15 1405 67
XM_017019006.1 XP_016874495.1 13 - 14 1298 67
XM_017019007.1 XP_016874496.1 13 - 14 1333 67
XM_017019008.1 XP_016874497.1 12 - 13 1200 67
XM_017019009.1 XP_016874498.1 13 - 14 1254 67
XM_017019010.1 XP_016874499.1 12 - 13 1226 67
XM_017019011.1 XP_016874500.1 15 - 16 1449 67
XM_017019012.1 XP_016874501.1 12 - 13 1236 67
XM_017019013.1 XP_016874502.1 12 - 13 1268 67
XM_017019014.1 XP_016874503.1 12 - 13 1303 67
XM_017019015.1 XP_016874504.1 12 - 13 1445 67
XM_017019016.1 XP_016874505.1 13 - 14 1464 67
XM_017019017.1 XP_016874506.1 12 - 13 1226 67
XM_017019018.1 XP_016874507.1 13 - 14 1294 67
XM_017019019.1 XP_016874508.1 14 - 15 1390 67
XM_017019020.1 XP_016874509.1 11 - 12 1140 67
XM_017019021.1 XP_016874510.1 12 - 13 1262 67
XM_017019022.1 XP_016874511.1 11 - 12 1172 67
XM_017019023.1 XP_016874512.1 11 - 12 1183 67
XM_017019024.1 XP_016874513.1 11 - 12 1172 67
XM_017019025.1 XP_016874514.1 12 - 13 1239 67
XM_017019026.1 XP_016874515.1 13 - 14 1279 67
XM_017019027.1 XP_016874516.1 12 - 13 1410 67
XM_017019028.1 XP_016874517.1 11 - 12 1172 67
XM_017019029.1 XP_016874518.1 11 - 12 1258 67
XM_017019030.1 XP_016874519.1 4 - 5 539 67
XM_017019031.1 XP_016874520.1 6 - 7 721 67
XM_017019032.1 XP_016874521.1 3 - 4 391 67
XM_017019033.1 XP_016874522.1 5 - 6 542 67
GenBank mRNA AB023219.1 - 13 - 14 1517 67
AK293394.1 - 3 - 4 412 67
AX775761.1 - 13 - 14 1517 67
AX775763.1 - 13 - 14 1559 67
BC041857.1 - 3 - 4 972 67
BC104995.1 - 1 - 2 167 67
BC112226.1 - 1 - 2 167 67
BC143605.1 - 1 - 2 167 67
BQ346058.1 - 1 - 2 67

View Full Product Details