Assay Design



Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 12 - 13 1341 75
XM_005268725.3 XP_005268782.1 14 - 15 1691 75
XM_011538036.1 XP_011536338.1 15 - 16 1531 75
XM_011538039.1 XP_011536341.1 14 - 15 1477 75
XM_011538040.1 XP_011536342.1 15 - 16 1531 75
XM_017019002.1 XP_016874491.1 14 - 15 1507 75
XM_017019008.1 XP_016874497.1 13 - 14 1381 75
XM_017019009.1 XP_016874498.1 14 - 15 1435 75
XM_017019012.1 XP_016874501.1 13 - 14 1417 75
XM_017019018.1 XP_016874507.1 14 - 15 1475 75
XM_017019019.1 XP_016874508.1 15 - 16 1571 75
XM_017019020.1 XP_016874509.1 12 - 13 1321 75
XM_017019025.1 XP_016874514.1 13 - 14 1420 75
XM_017019026.1 XP_016874515.1 14 - 15 1460 75
XM_017019027.1 XP_016874516.1 13 - 14 1591 75
XM_017019028.1 XP_016874517.1 12 - 13 1353 75
XM_017019029.1 XP_016874518.1 12 - 13 1439 75
XM_017019030.1 XP_016874519.1 5 - 6 720 75
XM_017019032.1 XP_016874521.1 4 - 5 572 75
XM_017019033.1 XP_016874522.1 6 - 7 723 75
GenBank mRNA AB023219.1 - 14 - 15 1698 75
AK293394.1 - 4 - 5 593 75
AX775761.1 - 14 - 15 1698 75
AX775763.1 - 14 - 15 1740 75
BC041857.1 - 4 - 5 1153 75
BC104995.1 - 2 - 3 348 75
BC112226.1 - 2 - 3 348 75

View Full Product Details