Assay Design



Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 4 - 5 460 132
XM_005268725.3 XP_005268782.1 6 - 7 810 132
XM_011538031.1 XP_011536333.1 6 - 7 554 132
XM_011538033.1 XP_011536335.1 6 - 7 554 132
XM_011538035.1 XP_011536337.1 6 - 7 554 132
XM_011538036.1 XP_011536338.1 6 - 7 554 132
XM_011538037.1 XP_011536339.1 5 - 6 530 132
XM_011538038.1 XP_011536340.1 6 - 7 554 132
XM_011538039.1 XP_011536341.1 6 - 7 554 132
XM_011538040.1 XP_011536342.1 6 - 7 554 132
XM_011538042.2 XP_011536344.1 7 - 8 650 132
XM_017018998.1 XP_016874487.1 5 - 6 530 132
XM_017018999.1 XP_016874488.1 7 - 8 691 132
XM_017019000.1 XP_016874489.1 6 - 7 554 132
XM_017019001.1 XP_016874490.1 6 - 7 764 132
XM_017019002.1 XP_016874491.1 5 - 6 530 132
XM_017019003.1 XP_016874492.1 6 - 7 554 132
XM_017019004.1 XP_016874493.1 6 - 7 554 132
XM_017019005.1 XP_016874494.1 6 - 7 609 132
XM_017019006.1 XP_016874495.1 5 - 6 502 132
XM_017019007.1 XP_016874496.1 5 - 6 537 132
XM_017019008.1 XP_016874497.1 6 - 7 554 132
XM_017019009.1 XP_016874498.1 6 - 7 554 132
XM_017019010.1 XP_016874499.1 5 - 6 526 132
XM_017019011.1 XP_016874500.1 8 - 9 707 132
XM_017019012.1 XP_016874501.1 6 - 7 554 132
XM_017019013.1 XP_016874502.1 5 - 6 526 132
XM_017019014.1 XP_016874503.1 5 - 6 561 132
XM_017019015.1 XP_016874504.1 4 - 5 649 132
XM_017019016.1 XP_016874505.1 6 - 7 764 132
XM_017019017.1 XP_016874506.1 5 - 6 526 132
XM_017019018.1 XP_016874507.1 5 - 6 498 132
XM_017019019.1 XP_016874508.1 7 - 8 648 132
XM_017019020.1 XP_016874509.1 6 - 7 554 132
XM_017019021.1 XP_016874510.1 6 - 7 616 132
XM_017019022.1 XP_016874511.1 5 - 6 526 132
XM_017019023.1 XP_016874512.1 5 - 6 537 132
XM_017019024.1 XP_016874513.1 5 - 6 526 132
XM_017019025.1 XP_016874514.1 5 - 6 503 132
XM_017019026.1 XP_016874515.1 7 - 8 633 132
XM_017019027.1 XP_016874516.1 6 - 7 764 132
XM_017019028.1 XP_016874517.1 5 - 6 526 132
XM_017019029.1 XP_016874518.1 6 - 7 672 132
GenBank mRNA AB023219.1 - 6 - 7 817 132
AX775761.1 - 6 - 7 817 132
AX775763.1 - 6 - 7 817 132

View Full Product Details