Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014925.3 | NP_055740.3 | 8 - 9 | 844 | 77 |
XM_005268725.3 | XP_005268782.1 | 10 - 11 | 1194 | 77 | |
XM_011538035.1 | XP_011536337.1 | 10 - 11 | 938 | 77 | |
XM_011538037.1 | XP_011536339.1 | 9 - 10 | 914 | 77 | |
XM_017019000.1 | XP_016874489.1 | 10 - 11 | 938 | 77 | |
XM_017019004.1 | XP_016874493.1 | 9 - 10 | 878 | 77 | |
XM_017019009.1 | XP_016874498.1 | 10 - 11 | 938 | 77 | |
XM_017019010.1 | XP_016874499.1 | 9 - 10 | 910 | 77 | |
XM_017019016.1 | XP_016874505.1 | 10 - 11 | 1148 | 77 | |
XM_017019017.1 | XP_016874506.1 | 9 - 10 | 910 | 77 | |
GenBank mRNA | AB023219.1 | - | 10 - 11 | 1201 | 77 |
AX775761.1 | - | 10 - 11 | 1201 | 77 |