Assay Design



Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 9 - 10 883 110
XM_005268725.3 XP_005268782.1 11 - 12 1233 110
XM_011538033.1 XP_011536335.1 12 - 13 1073 110
XM_011538035.1 XP_011536337.1 11 - 12 977 110
XM_011538036.1 XP_011536338.1 12 - 13 1073 110
XM_011538037.1 XP_011536339.1 10 - 11 953 110
XM_011538040.1 XP_011536342.1 12 - 13 1073 110
XM_011538042.2 XP_011536344.1 13 - 14 1169 110
XM_017018999.1 XP_016874488.1 13 - 14 1210 110
XM_017019000.1 XP_016874489.1 11 - 12 977 110
XM_017019001.1 XP_016874490.1 12 - 13 1283 110
XM_017019002.1 XP_016874491.1 11 - 12 1049 110
XM_017019004.1 XP_016874493.1 10 - 11 917 110
XM_017019005.1 XP_016874494.1 12 - 13 1128 110
XM_017019006.1 XP_016874495.1 11 - 12 1021 110
XM_017019007.1 XP_016874496.1 11 - 12 1056 110
XM_017019009.1 XP_016874498.1 11 - 12 977 110
XM_017019010.1 XP_016874499.1 10 - 11 949 110
XM_017019015.1 XP_016874504.1 10 - 11 1168 110
XM_017019016.1 XP_016874505.1 11 - 12 1187 110
XM_017019017.1 XP_016874506.1 10 - 11 949 110
XM_017019018.1 XP_016874507.1 11 - 12 1017 110
XM_017019025.1 XP_016874514.1 10 - 11 962 110
GenBank mRNA AB023219.1 - 11 - 12 1240 110
AX775761.1 - 11 - 12 1240 110

View Full Product Details