Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 7 - 8 | 1082 | 91 |
GenBank mRNA | AF439857.1 | - | 7 - 8 | 993 | 91 |
AK026048.1 | - | 1 - 2 | 203 | 91 | |
AK026840.1 | - | 7 - 8 | 992 | 91 | |
AK315545.1 | - | 7 - 8 | 1020 | 91 | |
BC016617.1 | - | 7 - 8 | 990 | 91 |