Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022916.4 | NP_075067.2 | 9 - 10 | 1280 | 77 |
GenBank mRNA | AF439857.1 | - | 9 - 10 | 1191 | 77 |
AK026048.1 | - | 3 - 4 | 401 | 77 | |
AK026840.1 | - | 9 - 10 | 1190 | 77 | |
AK315545.1 | - | 9 - 10 | 1218 | 77 | |
BC016617.1 | - | 9 - 10 | 1188 | 77 |