Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300985.1 | NP_001287914.1 | 7 - 8 | 969 | 86 |
NM_001300986.1 | NP_001287915.1 | 7 - 8 | 993 | 86 | |
NM_001300987.1 | NP_001287916.1 | 8 - 9 | 1125 | 86 | |
NM_001300988.1 | NP_001287917.1 | 7 - 8 | 969 | 86 | |
NM_001300989.1 | NP_001287918.1 | 7 - 8 | 969 | 86 | |
NM_001300990.1 | NP_001287919.1 | 6 - 7 | 893 | 86 | |
NM_021190.3 | NP_067013.1 | 7 - 8 | 969 | 86 | |
NR_125356.1 | 7 - 8 | 861 | 86 | ||
NR_125357.1 | 7 - 8 | 969 | 86 | ||
XM_011541874.2 | XP_011540176.1 | 7 - 8 | 802 | 86 | |
XM_011541875.2 | XP_011540177.1 | 7 - 8 | 815 | 86 | |
XM_011541876.2 | XP_011540178.1 | 7 - 8 | 832 | 86 | |
XM_017001951.1 | XP_016857440.1 | 6 - 7 | 832 | 86 | |
GenBank mRNA | AB051232.1 | - | 7 - 8 | 791 | 86 |
AB051233.1 | - | 7 - 8 | 735 | 86 | |
AF176085.1 | - | 7 - 8 | 761 | 86 | |
AF530580.1 | - | 7 - 8 | 764 | 86 | |
AF530581.1 | - | 7 - 8 | 764 | 86 | |
AK022716.1 | - | 2 - 3 | 275 | 86 | |
AK092216.1 | - | 6 - 7 | 833 | 86 | |
AK295384.1 | - | 7 - 8 | 782 | 86 | |
AK299755.1 | - | 6 - 7 | 578 | 86 | |
AK299897.1 | - | 8 - 9 | 906 | 86 | |
AK299922.1 | - | 7 - 8 | 807 | 86 | |
BC016582.2 | - | 7 - 8 | 738 | 86 |