Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005417.4 | NP_005408.1 | 9 - 10 | 1312 | 79 |
NM_198291.2 | NP_938033.1 | 9 - 10 | 1237 | 79 | |
XM_011529013.2 | XP_011527315.1 | 10 - 11 | 1581 | 79 | |
XM_017028024.1 | XP_016883513.1 | 11 - 12 | 1605 | 79 | |
XM_017028025.1 | XP_016883514.1 | 10 - 11 | 1486 | 79 | |
XM_017028026.1 | XP_016883515.1 | 10 - 11 | 1325 | 79 | |
XM_017028027.1 | XP_016883516.1 | 8 - 9 | 902 | 79 | |
GenBank mRNA | AK024281.1 | - | 2 - 3 | 509 | 79 |
BC011566.1 | - | 7 - 8 | 902 | 79 | |
BC051270.1 | - | 7 - 8 | 965 | 79 |