Assay Design



Gene Transcripts

Gene Symbol
TTLL9
Entrez Gene ID
164395
Gene Name
tubulin tyrosine ligase like 9
Gene Aliases
C20orf125
Chromosome Location
Chr.20: 31870702 - 31944959 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006723723.2 XP_006723786.1 6 - 7 589 89
XM_006723724.2 XP_006723787.1 8 - 9 830 89
XM_011528617.1 XP_011526919.1 9 - 10 793 89
XM_011528618.1 XP_011526920.1 9 - 10 957 89
XM_011528619.1 XP_011526921.1 10 - 11 855 89
XM_011528620.2 XP_011526922.1 10 - 11 852 89
XM_011528621.1 XP_011526923.1 10 - 11 870 89
XM_011528622.1 XP_011526924.1 10 - 11 845 89
XM_011528623.2 XP_011526925.1 8 - 9 726 89
XM_011528624.1 XP_011526926.1 8 - 9 724 89
XM_011528625.1 XP_011526927.1 9 - 10 793 89
XM_011528626.1 XP_011526928.1 9 - 10 791 89
XM_011528627.2 XP_011526929.1 9 - 10 788 89
XM_011528628.1 XP_011526930.1 8 - 9 716 89
XM_011528629.1 XP_011526931.1 8 - 9 724 89
XM_011528630.1 XP_011526932.1 7 - 8 658 89
XM_011528631.1 XP_011526933.1 8 - 9 724 89
XM_011528632.1 XP_011526934.1 9 - 10 793 89
XM_011528633.1 XP_011526935.1 9 - 10 793 89
XM_011528634.1 XP_011526936.1 9 - 10 793 89
XM_011528635.1 XP_011526937.1 9 - 10 793 89
XM_011528638.1 XP_011526940.1 9 - 10 793 89
XM_011528639.1 XP_011526941.1 9 - 10 793 89
XM_011528640.1 XP_011526942.1 9 - 10 793 89
XM_011528646.1 XP_011526948.1 9 - 10 913 89
XM_017027694.1 XP_016883183.1 6 - 7 589 89
XM_017027695.1 XP_016883184.1 9 - 10 793 89
XM_017027697.1 XP_016883186.1 9 - 10 793 89
XM_017027698.1 XP_016883187.1 9 - 10 793 89
XM_017027699.1 XP_016883188.1 8 - 9 976 89
XM_017027701.1 XP_016883190.1 9 - 10 793 89
XM_017027703.1 XP_016883192.1 8 - 9 724 89
GenBank mRNA AK302360.1 - 6 - 7 591 89
AK303963.1 - 8 - 9 595 89
BC104024.1 - 9 - 10 913 89
BC104025.1 - 8 - 9 830 89

View Full Product Details