Assay Design



Gene Transcripts

Gene Symbol
PLCB4
Entrez Gene ID
5332
Gene Name
phospholipase C beta 4
Gene Aliases
ARCND2, PI-PLC
Chromosome Location
Chr.20: 9068710 - 9480816 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172646.1 NP_001166117.1 19 - 20 1839 70
XM_005260724.2 XP_005260781.1 19 - 20 1880 70
XM_005260728.2 XP_005260785.1 10 - 11 1047 70
XM_006723568.3 XP_006723631.1 20 - 21 1862 70
XM_006723569.2 XP_006723632.1 19 - 20 1960 70
XM_011529254.2 XP_011527556.1 12 - 13 4982 70
XM_017027882.1 XP_016883371.1 15 - 16 1464 70
XM_017027884.1 XP_016883373.1 17 - 18 1914 70
XM_017027885.1 XP_016883374.1 17 - 18 1591 70
GenBank mRNA AK122699.1 - 19 - 20 1803 70
AK307837.1 - 19 - 20 1840 70

View Full Product Details