Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256135.1 | NP_001243064.1 | 11 - 12 | 1360 | 158 |
NM_001316.3 | NP_001307.2 | 12 - 13 | 1528 | 158 | |
NR_045796.1 | 9 - 10 | 1043 | 158 | ||
XM_011528599.2 | XP_011526901.1 | 12 - 13 | 1462 | 158 | |
GenBank mRNA | AF053641.1 | - | 12 - 13 | 1462 | 158 |
AK297270.1 | - | 8 - 9 | 829 | 158 | |
AK298477.1 | - | 5 - 6 | 524 | 158 | |
AK300589.1 | - | 9 - 10 | 978 | 158 | |
AK307965.1 | - | 5 - 6 | 474 | 158 | |
AK312306.1 | - | 12 - 13 | 1459 | 158 | |
BC108309.1 | - | 12 - 13 | 1474 | 158 | |
BC109313.2 | - | 12 - 13 | 1441 | 158 | |
BC109314.2 | - | 12 - 13 | 1441 | 158 | |
EF426455.1 | - | 10 - 11 | 1171 | 158 | |
U33286.1 | - | 12 - 13 | 1462 | 158 |