Assay Design



Gene Transcripts

Gene Symbol
CSE1L
Entrez Gene ID
1434
Gene Name
chromosome segregation 1 like
Gene Aliases
CAS, CSE1, XPO2
Chromosome Location
Chr.20: 49046246 - 49096960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256135.1 NP_001243064.1 22 - 23 2611 147
NM_001316.3 NP_001307.2 23 - 24 2779 147
NR_045796.1 20 - 21 2294 147
XM_011528599.2 XP_011526901.1 23 - 24 2713 147
GenBank mRNA AF053642.1 - 4 - 5 389 147
AK098618.1 - 9 - 10 1059 147
AK225610.1 - 15 - 16 1652 147
AK297270.1 - 19 - 20 2080 147
AK298477.1 - 16 - 17 1775 147
AK299549.1 - 14 - 15 1580 147
AK300589.1 - 20 - 21 2229 147
AK312306.1 - 23 - 24 2710 147
BC108309.1 - 23 - 24 2725 147
BC109313.2 - 23 - 24 2692 147
BC109314.2 - 23 - 24 2692 147
EF426455.1 - 21 - 22 2422 147
U33286.1 - 23 - 24 2713 147

View Full Product Details