Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002915.3 | NP_002906.1 | 3 - 4 | 403 | 101 |
NM_181558.2 | NP_853536.2 | 3 - 4 | 403 | 101 | |
XM_011535172.2 | XP_011533474.1 | 3 - 4 | 423 | 101 | |
XM_011535173.2 | XP_011533475.1 | 3 - 4 | 423 | 101 | |
XM_011535174.2 | XP_011533476.1 | 3 - 4 | 423 | 101 | |
XM_011535175.1 | XP_011533477.1 | 3 - 4 | 423 | 101 | |
XM_017020680.1 | XP_016876169.1 | 3 - 4 | 423 | 101 | |
XM_017020681.1 | XP_016876170.1 | 3 - 4 | 423 | 101 | |
XM_017020682.1 | XP_016876171.1 | 3 - 4 | 423 | 101 | |
XM_017020683.1 | XP_016876172.1 | 3 - 4 | 423 | 101 | |
GenBank mRNA | AK222572.1 | - | 3 - 4 | 330 | 101 |
AK296527.1 | - | 3 - 4 | 344 | 101 | |
AK312544.1 | - | 3 - 4 | 320 | 101 | |
AL596753.1 | - | 3 - 4 | 431 | 101 | |
BC000149.2 | - | 3 - 4 | 334 | 101 | |
BT019668.1 | - | 3 - 4 | 293 | 101 | |
CX786577.1 | - | 2 - 3 | 145 | 101 | |
DA943219.1 | - | 3 - 4 | 403 | 101 | |
L07541.1 | - | 3 - 4 | 302 | 101 |