Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002199.3 | NP_002190.2 | 3 - 4 | 388 | 123 |
GenBank mRNA | AK312953.1 | - | 3 - 4 | 366 | 123 |
BC015803.2 | - | 2 - 3 | 195 | 123 | |
BP200998.1 | - | 3 - 4 | 388 | 123 | |
BT007264.1 | - | 2 - 3 | 188 | 123 | |
CR457077.1 | - | 2 - 3 | 188 | 123 | |
X15949.1 | - | 3 - 4 | 286 | 123 |