Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005245.3 | NP_005236.2 | 19 - 20 | 11544 | 66 |
XM_005262834.2 | XP_005262891.1 | 19 - 20 | 11936 | 66 | |
XM_005262835.2 | XP_005262892.1 | 19 - 20 | 11561 | 66 | |
XM_006714139.2 | XP_006714202.1 | 19 - 20 | 11936 | 66 | |
GenBank mRNA | X87241.1 | - | 19 - 20 | 11548 | 66 |