Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308088.1 | NP_001295017.1 | 3 - 4 | 327 | 150 |
XM_011543454.2 | XP_011541756.1 | 3 - 4 | 574 | 150 | |
GenBank mRNA | BC038838.1 | - | 2 - 3 | 186 | 150 |
BM728431.1 | - | 3 - 4 | 327 | 150 |