Assay Design



Gene Transcripts

Gene Symbol
SASH1
Entrez Gene ID
23328
Gene Name
SAM and SH3 domain containing 1
Gene Aliases
SH3D6A, dJ323M4, dJ323M4.1
Chromosome Location
Chr.6: 148193440 - 148552049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015278.3 NP_056093.3 19 - 20 3955 67
XM_005266875.1 XP_005266932.1 12 - 13 3181 67
XM_011535640.1 XP_011533942.1 19 - 20 3398 67
XM_017010598.1 XP_016866087.1 19 - 20 4676 67
XM_017010599.1 XP_016866088.1 19 - 20 3999 67
XM_017010600.1 XP_016866089.1 19 - 20 3887 67
XM_017010601.1 XP_016866090.1 18 - 19 4101 67
XM_017010602.1 XP_016866091.1 17 - 18 3798 67
XM_017010603.1 XP_016866092.1 17 - 18 3737 67
XM_017010604.1 XP_016866093.1 16 - 17 3646 67
XM_017010605.1 XP_016866094.1 16 - 17 3612 67
XM_017010606.1 XP_016866095.1 12 - 13 3670 67
XM_017010607.1 XP_016866096.1 20 - 21 3578 67
GenBank mRNA AB018333.1 - 19 - 20 3698 67
AJ507735.1 - 19 - 20 3486 67
AK024403.1 - 13 - 14 67
AK025495.1 - 19 - 20 3955 67
AK074249.1 - 9 - 10 2240 67
BC028303.1 - 2 - 3 466 67
BC063279.1 - 19 - 20 3424 67

View Full Product Details