Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015278.3 | NP_056093.3 | 6 - 7 | 992 | 88 |
XM_011535640.1 | XP_011533942.1 | 6 - 7 | 435 | 88 | |
XM_017010598.1 | XP_016866087.1 | 6 - 7 | 1224 | 88 | |
XM_017010599.1 | XP_016866088.1 | 6 - 7 | 547 | 88 | |
XM_017010600.1 | XP_016866089.1 | 6 - 7 | 435 | 88 | |
XM_017010601.1 | XP_016866090.1 | 5 - 6 | 649 | 88 | |
XM_017010602.1 | XP_016866091.1 | 4 - 5 | 346 | 88 | |
XM_017010603.1 | XP_016866092.1 | 4 - 5 | 285 | 88 | |
XM_017010604.1 | XP_016866093.1 | 3 - 4 | 194 | 88 | |
XM_017010605.1 | XP_016866094.1 | 3 - 4 | 160 | 88 | |
XM_017010607.1 | XP_016866096.1 | 7 - 8 | 615 | 88 | |
GenBank mRNA | AB018333.1 | - | 6 - 7 | 735 | 88 |
AJ507735.1 | - | 6 - 7 | 523 | 88 | |
AK025495.1 | - | 6 - 7 | 992 | 88 | |
BC063279.1 | - | 6 - 7 | 461 | 88 |