Assay Design



Gene Transcripts

Gene Symbol
REV3L
Entrez Gene ID
5980
Gene Name
REV3 like, DNA directed polymerase zeta catalytic subunit
Gene Aliases
POLZ, REV3
Chromosome Location
Chr.6: 111299031 - 111483715 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286432.1 NP_001273361.1 3 - 4 725 154
XM_011536031.2 XP_011534333.1 2 - 3 816 154
XM_011536032.2 XP_011534334.1 2 - 3 5406 154
XM_017011152.1 XP_016866641.1 2 - 3 816 154
XM_017011153.1 XP_016866642.1 2 - 3 359 154
XM_017011154.1 XP_016866643.1 2 - 3 244 154
GenBank mRNA AF078695.1 - 3 - 4 727 154
AF179428.1 - 2 - 3 298 154
AK310949.1 - 3 - 4 721 154

View Full Product Details