Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308237.1 | NP_001295166.1 | 10 - 11 | 1194 | 111 |
NM_015534.5 | NP_056349.1 | 11 - 12 | 2658 | 111 | |
XM_005270725.3 | XP_005270782.1 | 11 - 12 | 2733 | 111 | |
XM_005270726.3 | XP_005270783.1 | 10 - 11 | 2634 | 111 | |
XM_005270727.3 | XP_005270784.1 | 11 - 12 | 2730 | 111 | |
XM_005270729.4 | XP_005270786.1 | 9 - 10 | 1144 | 111 | |
XM_017000970.1 | XP_016856459.1 | 10 - 11 | 2631 | 111 | |
XM_017000972.1 | XP_016856461.1 | 10 - 11 | 1162 | 111 | |
GenBank mRNA | AK074119.1 | - | 6 - 7 | 3073 | 111 |
AL080063.1 | - | 3 - 4 | 222 | 111 | |
BC035079.2 | - | 10 - 11 | 1186 | 111 | |
BC035397.1 | - | 7 - 8 | 1326 | 111 | |
BC035818.2 | - | 7 - 8 | 2233 | 111 | |
BX640658.1 | - | 7 - 8 | 746 | 111 | |
BX640766.1 | - | 7 - 8 | 1932 | 111 | |
BX641001.1 | - | 11 - 12 | 2659 | 111 | |
BX641096.1 | - | 11 - 12 | 2642 | 109 |