Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153042.3 | NP_694587.3 | 5 - 6 | 550 | 140 |
XM_005248925.1 | XP_005248982.1 | 5 - 6 | 550 | 140 | |
XM_005248926.1 | XP_005248983.1 | 5 - 6 | 550 | 140 | |
XM_005248928.1 | XP_005248985.1 | 5 - 6 | 550 | 140 | |
XM_011514385.1 | XP_011512687.1 | 5 - 6 | 550 | 140 | |
XM_011514386.1 | XP_011512688.1 | 5 - 6 | 550 | 140 | |
XM_017010440.1 | XP_016865929.1 | 5 - 6 | 550 | 140 | |
XM_017010441.1 | XP_016865930.1 | 5 - 6 | 550 | 140 | |
XM_017010443.1 | XP_016865932.1 | 4 - 5 | 343 | 140 | |
XM_017010445.1 | XP_016865934.1 | 5 - 6 | 550 | 140 | |
XM_017010446.1 | XP_016865935.1 | 5 - 6 | 550 | 140 | |
GenBank mRNA | AK125318.1 | - | 5 - 6 | 486 | 140 |