Assay Design



Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 14 - 15 1892 140
GenBank mRNA AF385437.1 - 14 - 15 1892 140
AK074202.1 - 8 - 9 942 140
AK095312.1 - 14 - 15 1750 140
AL832494.1 - 13 - 14 1694 140
BC133025.1 - 14 - 15 1852 140
BC136517.1 - 14 - 15 1852 140
CR749211.1 - 9 - 10 1092 140

View Full Product Details