Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270484.1 | NP_001257413.1 | 2 - 3 | 627 | 70 |
NM_003181.3 | NP_003172.1 | 2 - 3 | 720 | 70 | |
XM_011536080.2 | XP_011534382.1 | 1 - 2 | 487 | 70 | |
GenBank mRNA | AJ001699.1 | - | 1 - 2 | 360 | 70 |
BC039877.1 | - | 1 - 2 | 487 | 70 | |
BC098425.1 | - | 2 - 3 | 600 | 70 | |
BE785653.1 | - | 2 - 3 | 626 | 70 | |
BU190437.1 | - | 2 - 3 | 671 | 70 |