Assay Design



Gene Transcripts

Gene Symbol
SLC4A10
Entrez Gene ID
57282
Gene Name
solute carrier family 4 member 10
Gene Aliases
NBCn2, NCBE
Chromosome Location
Chr.2: 161624335 - 161985276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178015.1 NP_001171486.1 19 - 20 2725 74
NM_001178016.1 NP_001171487.1 19 - 20 2771 74
NM_022058.3 NP_071341.2 18 - 19 2635 74
XM_005246694.4 XP_005246751.1 20 - 21 2780 74
XM_005246695.4 XP_005246752.1 19 - 20 2690 74
XM_011511508.2 XP_011509810.1 19 - 20 2568 74
XM_011511509.2 XP_011509811.1 19 - 20 2565 74
XM_011511510.2 XP_011509812.1 19 - 20 2568 74
XM_011511511.2 XP_011509813.1 18 - 19 2478 74
XM_011511512.2 XP_011509814.1 18 - 19 2475 74
XM_011511513.2 XP_011509815.1 22 - 23 3057 74
XM_017004541.1 XP_016860030.1 19 - 20 2565 74
XM_017004542.1 XP_016860031.1 18 - 19 2475 74
XM_017004543.1 XP_016860032.1 18 - 19 2478 74
XM_017004544.1 XP_016860033.1 18 - 19 2475 74
XM_017004545.1 XP_016860034.1 21 - 22 2966 74
XM_017004546.1 XP_016860035.1 21 - 22 2967 74
XM_017004547.1 XP_016860036.1 20 - 21 2780 74
XM_017004548.1 XP_016860037.1 20 - 21 2754 74
XM_017004549.1 XP_016860038.1 20 - 21 2842 74
XM_017004550.1 XP_016860039.1 19 - 20 2687 74
XM_017004551.1 XP_016860040.1 15 - 16 2063 74
XM_017004552.1 XP_016860041.1 19 - 20 2584 74
XM_017004553.1 XP_016860042.1 19 - 20 2584 74
XM_017004554.1 XP_016860043.1 9 - 10 1767 74
GenBank mRNA AB040457.1 - 18 - 19 2543 74
AK293793.1 - 19 - 20 2684 74
BC136269.1 - 18 - 19 2521 74
BC143714.1 - 19 - 20 2611 74

View Full Product Details