Assay Design



Gene Transcripts

Gene Symbol
SLC4A10
Entrez Gene ID
57282
Gene Name
solute carrier family 4 member 10
Gene Aliases
NBCn2, NCBE
Chromosome Location
Chr.2: 161624335 - 161985276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178015.1 NP_001171486.1 3 - 4 456 130
NM_001178016.1 NP_001171487.1 4 - 5 592 130
NM_022058.3 NP_071341.2 3 - 4 456 130
XM_005246694.4 XP_005246751.1 4 - 5 511 130
XM_005246695.4 XP_005246752.1 4 - 5 511 130
XM_011511508.2 XP_011509810.1 3 - 4 299 130
XM_011511509.2 XP_011509811.1 3 - 4 299 130
XM_011511510.2 XP_011509812.1 3 - 4 299 130
XM_011511511.2 XP_011509813.1 3 - 4 299 130
XM_011511512.2 XP_011509814.1 3 - 4 299 130
XM_011511513.2 XP_011509815.1 6 - 7 788 130
XM_017004541.1 XP_016860030.1 3 - 4 299 130
XM_017004542.1 XP_016860031.1 3 - 4 299 130
XM_017004543.1 XP_016860032.1 3 - 4 299 130
XM_017004544.1 XP_016860033.1 3 - 4 299 130
XM_017004545.1 XP_016860034.1 5 - 6 697 130
XM_017004546.1 XP_016860035.1 6 - 7 788 130
XM_017004547.1 XP_016860036.1 4 - 5 511 130
XM_017004548.1 XP_016860037.1 4 - 5 485 130
XM_017004549.1 XP_016860038.1 5 - 6 663 130
XM_017004550.1 XP_016860039.1 4 - 5 511 130
XM_017004552.1 XP_016860041.1 3 - 4 299 130
XM_017004553.1 XP_016860042.1 3 - 4 299 130
GenBank mRNA AB040457.1 - 3 - 4 364 130
AK293793.1 - 4 - 5 505 130
AK294767.1 - 3 - 4 393 130
AK308529.1 - 6 - 7 788 130
AK309839.1 - 4 - 5 511 130
AY376402.1 - 3 - 4 272 130
BC136269.1 - 3 - 4 342 130
BC143714.1 - 3 - 4 342 130
DC337667.1 - 3 - 4 457 130

View Full Product Details