Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001410.2 | NP_001001410.1 | 1 - 2 | 182 | 121 |
GenBank mRNA | BC013943.1 | - | 1 - 2 | 186 | 121 |
BC016699.1 | - | 1 - 2 | 186 | 121 | |
BC017325.2 | - | 1 - 2 | 166 | 121 |