Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016941.3 | NP_058637.1 | 2 - 3 | 413 | 77 |
NM_203486.2 | NP_982353.1 | 2 - 3 | 413 | 77 | |
GenBank mRNA | AK075302.1 | - | 2 - 3 | 383 | 77 |
AK308460.1 | - | 2 - 3 | 435 | 77 | |
BC000218.2 | - | 2 - 3 | 362 | 77 | |
DB035825.1 | - | 2 - 3 | 413 | 77 |