Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013233.2 | NP_037365.2 | 12 - 13 | 1408 | 134 |
XM_017003813.1 | XP_016859302.1 | 12 - 13 | 1354 | 134 | |
XM_017003814.1 | XP_016859303.1 | 12 - 13 | 1354 | 134 | |
XM_017003815.1 | XP_016859304.1 | 12 - 13 | 2694 | 134 | |
XM_017003817.1 | XP_016859306.1 | 9 - 10 | 767 | 134 | |
GenBank mRNA | AF017635.1 | - | 6 - 7 | 440 | 134 |
AF030403.1 | - | 12 - 13 | 1500 | 134 | |
AF099989.1 | - | 12 - 13 | 1427 | 134 | |
BX647113.1 | - | 3 - 4 | 349 | 134 | |
KJ534949.1 | - | 13 - 14 | 1191 | 134 | |
KJ534951.1 | - | 13 - 14 | 1182 | 134 | |
KJ535059.1 | - | 13 - 14 | 1171 | 134 |