Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013233.2 | NP_037365.2 | 5 - 6 | 793 | 129 |
XM_005246465.2 | XP_005246522.1 | 5 - 6 | 739 | 129 | |
XM_017003813.1 | XP_016859302.1 | 5 - 6 | 739 | 129 | |
XM_017003814.1 | XP_016859303.1 | 5 - 6 | 739 | 129 | |
XM_017003815.1 | XP_016859304.1 | 5 - 6 | 2079 | 129 | |
XM_017003816.1 | XP_016859305.1 | 5 - 6 | 739 | 129 | |
XM_017003817.1 | XP_016859306.1 | 2 - 3 | 152 | 129 | |
GenBank mRNA | AF030403.1 | - | 5 - 6 | 885 | 129 |
AF099989.1 | - | 5 - 6 | 812 | 129 | |
KJ534948.1 | - | 6 - 7 | 576 | 129 | |
KJ534949.1 | - | 6 - 7 | 576 | 129 | |
KJ534950.1 | - | 6 - 7 | 588 | 129 | |
KJ534951.1 | - | 6 - 7 | 567 | 129 | |
KJ535059.1 | - | 6 - 7 | 556 | 129 |