Assay Design



Gene Transcripts

Gene Symbol
KBTBD8
Entrez Gene ID
84541
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
TA-KRP, TAKRP
Chromosome Location
Chr.3: 66998303 - 67011210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032505.2 NP_115894.2 1 - 2 69 83
GenBank mRNA AK096640.1 - 1 - 2 65 83
DA246470.1 - 1 - 2 65 83
DB065239.1 - 1 - 2 69 83

View Full Product Details