Assay Design



Gene Transcripts

Gene Symbol
KBTBD8
Entrez Gene ID
84541
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
TA-KRP, TAKRP
Chromosome Location
Chr.3: 66998303 - 67011210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032505.2 NP_115894.2 2 - 3 273 94
GenBank mRNA AF385438.1 - 1 - 2 206 94
AK096640.1 - 2 - 3 269 94
BC117487.1 - 1 - 2 170 94
DA246470.1 - 2 - 3 269 94
DB065239.1 - 2 - 3 274 94

View Full Product Details