Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251888.1 | NP_001238817.1 | 1 - 2 | 199 | 115 |
NM_024083.3 | NP_076988.1 | 1 - 2 | 199 | 115 | |
XM_011523601.2 | XP_011521903.1 | 1 - 2 | 199 | 115 | |
XM_011523602.2 | XP_011521904.1 | 1 - 2 | 199 | 115 | |
XM_017025036.1 | XP_016880525.1 | 1 - 2 | 199 | 115 | |
XM_017025037.1 | XP_016880526.1 | 1 - 2 | 199 | 115 | |
XM_017025042.1 | XP_016880531.1 | 1 - 2 | 199 | 115 | |
GenBank mRNA | AF324219.2 | - | 1 - 2 | 124 | 115 |
AK057403.1 | - | 1 - 2 | 199 | 115 | |
AK226086.1 | - | 1 - 2 | 72 | 115 | |
AK290624.1 | - | 1 - 2 | 115 | 115 | |
BC018722.1 | - | 1 - 2 | 104 | 115 |