Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251888.1 | NP_001238817.1 | 8 - 9 | 1188 | 91 |
NM_024083.3 | NP_076988.1 | 8 - 9 | 1188 | 91 | |
NR_045351.1 | 7 - 8 | 1132 | 91 | ||
XM_011523601.2 | XP_011521903.1 | 8 - 9 | 1188 | 91 | |
XM_011523602.2 | XP_011521904.1 | 8 - 9 | 1188 | 91 | |
XM_017025036.1 | XP_016880525.1 | 8 - 9 | 1188 | 91 | |
XM_017025037.1 | XP_016880526.1 | 8 - 9 | 1188 | 91 | |
XM_017025038.1 | XP_016880527.1 | 9 - 10 | 1199 | 91 | |
XM_017025039.1 | XP_016880528.1 | 9 - 10 | 1116 | 91 | |
XM_017025040.1 | XP_016880529.1 | 8 - 9 | 1053 | 91 | |
XM_017025041.1 | XP_016880530.1 | 7 - 8 | 1056 | 91 | |
XM_017025042.1 | XP_016880531.1 | 8 - 9 | 1188 | 91 | |
XM_017025043.1 | XP_016880532.1 | 7 - 8 | 1056 | 91 | |
XM_017025044.1 | XP_016880533.1 | 9 - 10 | 1199 | 91 | |
GenBank mRNA | AF324219.2 | - | 8 - 9 | 1113 | 91 |
AK057403.1 | - | 8 - 9 | 1188 | 91 | |
AK057851.1 | - | 7 - 8 | 1040 | 91 | |
AK090682.1 | - | 1 - 2 | 2553 | 91 | |
AK126967.1 | - | 1 - 2 | 4848 | 91 | |
AK127159.1 | - | 1 - 2 | 1760 | 91 | |
AK226086.1 | - | 8 - 9 | 1061 | 91 | |
AK290624.1 | - | 8 - 9 | 1104 | 91 | |
BC006152.1 | - | 8 - 9 | 1143 | 91 | |
BC018722.1 | - | 8 - 9 | 1093 | 91 |