Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164623.1 | NP_001158095.1 | 2 - 3 | 255 | 113 |
NM_012154.3 | NP_036286.2 | 2 - 3 | 255 | 113 | |
XM_011516965.2 | XP_011515267.1 | 2 - 3 | 3331 | 113 | |
XM_011516966.2 | XP_011515268.1 | 2 - 3 | 1072 | 113 | |
XM_011516968.2 | XP_011515270.2 | 2 - 3 | 406 | 113 | |
GenBank mRNA | AA909234.1 | - | 2 - 3 | 252 | 113 |
AI468419.1 | - | 2 - 3 | 255 | 113 | |
AY077717.1 | - | - | 193 | 113 |