Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004073.3 | NP_004064.2 | 11 - 12 | 1588 | 114 |
GenBank mRNA | AB209875.1 | - | 12 - 13 | 1275 | 114 |
BC013899.2 | - | 11 - 12 | 1449 | 114 | |
U56998.1 | - | 11 - 12 | 1268 | 114 |