Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078175.2 | NP_001071643.1 | 11 - 12 | 1270 | 79 |
NM_001078177.1 | NP_001071645.1 | 11 - 12 | 1465 | 79 | |
NM_001304462.1 | NP_001291391.1 | 11 - 12 | 1403 | 79 | |
NM_001304463.1 | NP_001291392.1 | 10 - 11 | 1163 | 79 | |
NM_001304465.1 | NP_001291394.1 | 10 - 11 | 1306 | 79 | |
NM_001304466.1 | NP_001291395.1 | 10 - 11 | 1303 | 79 | |
XM_005248876.4 | XP_005248933.1 | 10 - 11 | 1128 | 79 | |
XM_005248878.3 | XP_005248935.1 | 11 - 12 | 1300 | 79 | |
XM_005248879.3 | XP_005248936.1 | 11 - 12 | 1234 | 79 | |
XM_005248880.3 | XP_005248937.1 | 11 - 12 | 1211 | 79 | |
XM_005248881.3 | XP_005248938.1 | 10 - 11 | 1308 | 79 | |
XM_005248882.3 | XP_005248939.1 | 10 - 11 | 1251 | 79 | |
XM_011514341.2 | XP_011512643.1 | 11 - 12 | 1324 | 79 | |
GenBank mRNA | AF079117.1 | - | 10 - 11 | 1076 | 79 |
AK090491.1 | - | 11 - 12 | 1272 | 79 | |
AK090615.1 | - | 11 - 12 | 1291 | 79 | |
AX775997.1 | - | 10 - 11 | 1152 | 79 | |
BC001382.2 | - | 10 - 11 | 1076 | 79 | |
BC008954.2 | - | 11 - 12 | 1149 | 79 | |
U81375.1 | - | 10 - 11 | 1152 | 79 |