Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078175.2 | NP_001071643.1 | 4 - 5 | 406 | 98 |
NM_001078177.1 | NP_001071645.1 | 4 - 5 | 601 | 98 | |
NM_001304462.1 | NP_001291391.1 | 4 - 5 | 539 | 98 | |
NM_001304463.1 | NP_001291392.1 | 3 - 4 | 299 | 98 | |
NM_001304465.1 | NP_001291394.1 | 3 - 4 | 442 | 98 | |
NM_001304466.1 | NP_001291395.1 | 3 - 4 | 439 | 98 | |
XM_005248876.4 | XP_005248933.1 | 3 - 4 | 264 | 98 | |
XM_005248878.3 | XP_005248935.1 | 4 - 5 | 436 | 98 | |
XM_005248879.3 | XP_005248936.1 | 4 - 5 | 370 | 98 | |
XM_005248880.3 | XP_005248937.1 | 4 - 5 | 347 | 98 | |
XM_005248881.3 | XP_005248938.1 | 3 - 4 | 444 | 98 | |
XM_005248882.3 | XP_005248939.1 | 3 - 4 | 387 | 98 | |
XM_011514341.2 | XP_011512643.1 | 4 - 5 | 460 | 98 | |
GenBank mRNA | AB490708.1 | - | 4 - 5 | 395 | 98 |
AB490709.1 | - | 4 - 5 | 297 | 98 | |
AF079117.1 | - | 3 - 4 | 212 | 98 | |
AK090491.1 | - | 4 - 5 | 408 | 98 | |
AK090615.1 | - | 4 - 5 | 427 | 98 | |
AK293558.1 | - | 3 - 4 | 208 | 98 | |
AK304285.1 | - | 2 - 3 | 228 | 98 | |
AX775997.1 | - | 3 - 4 | 288 | 98 | |
BC001382.2 | - | 3 - 4 | 212 | 98 | |
BC008954.2 | - | 4 - 5 | 285 | 98 | |
CB996332.1 | - | 4 - 5 | 378 | 98 | |
DA680982.1 | - | 3 - 4 | 439 | 98 | |
DC396448.1 | - | 3 - 4 | 299 | 98 | |
U81375.1 | - | 3 - 4 | 288 | 98 |