Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004977.2 | NP_004968.2 | 3 - 4 | 2465 | 98 |
XM_006723203.2 | XP_006723266.1 | 3 - 4 | 2170 | 98 | |
XM_011526926.1 | XP_011525228.1 | 3 - 4 | 2170 | 98 | |
GenBank mRNA | AF055989.1 | - | 3 - 4 | 2465 | 98 |