Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114377.1 | NP_001107849.1 | 8 - 9 | 1047 | 107 |
NM_014009.3 | NP_054728.2 | 9 - 10 | 1152 | 107 | |
XM_006724533.2 | XP_006724596.2 | 8 - 9 | 1327 | 107 | |
XM_011543916.2 | XP_011542218.1 | 9 - 10 | 2577 | 107 | |
XM_017029565.1 | XP_016885054.1 | 10 - 11 | 2613 | 107 | |
XM_017029566.1 | XP_016885055.1 | 9 - 10 | 2577 | 107 | |
XM_017029567.1 | XP_016885056.1 | 8 - 9 | 972 | 107 | |
GenBank mRNA | AF277993.1 | - | 9 - 10 | 1152 | 107 |
AJ005891.1 | - | 8 - 9 | 814 | 107 | |
AK292052.1 | - | 9 - 10 | 1036 | 107 | |
AK299988.1 | - | 8 - 9 | 931 | 107 | |
BC113401.1 | - | 8 - 9 | 994 | 107 | |
BC113403.1 | - | 8 - 9 | 994 | 107 | |
BC143785.1 | - | 7 - 8 | 889 | 107 | |
BC143786.1 | - | 7 - 8 | 1063 | 107 | |
DQ010327.1 | - | 7 - 8 | 859 | 107 | |
EF534714.1 | - | 8 - 9 | 964 | 107 | |
EU855812.1 | - | 7 - 8 | 883 | 107 |