Assay Design
Gene Transcripts
- Gene Symbol
- SMARCD1
- Entrez Gene ID
-
6602
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
- Gene Aliases
-
BAF60A, CRACD1, Rsc6p
- Chromosome Location
-
Chr.12: 50085200 - 50100711 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139071.2 |
NP_620710.2 |
10 - 11 |
1442 |
70 |
GenBank mRNA |
U66617.1 |
- |
10 - 11 |
1420 |
70 |
View Full Product Details