Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003239.3 | NP_003230.1 | 3 - 4 | 1774 | 92 |
GenBank mRNA | AK313564.1 | - | 3 - 4 | 855 | 92 |
BC018503.1 | - | 3 - 4 | 745 | 92 | |
BT007287.1 | - | 3 - 4 | 645 | 92 | |
EF560714.1 | - | 3 - 4 | 758 | 92 | |
J03241.1 | - | 3 - 4 | 907 | 92 | |
X14149.1 | - | 3 - 4 | 898 | 92 |