Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193268.1 | NP_001180197.1 | 15 - 16 | 1928 | 110 |
NM_001193269.1 | NP_001180198.1 | 15 - 16 | 1832 | 110 | |
NM_012155.2 | NP_036287.1 | 12 - 13 | 1293 | 110 | |
NR_034098.1 | 10 - 11 | 1013 | 110 | ||
XM_006723125.2 | XP_006723188.1 | 15 - 16 | 2009 | 110 | |
XM_006723126.2 | XP_006723189.1 | 15 - 16 | 1817 | 110 | |
XM_011526694.2 | XP_011524996.1 | 14 - 15 | 1916 | 110 | |
XM_011526695.2 | XP_011524997.1 | 14 - 15 | 1877 | 110 | |
XM_011526698.2 | XP_011525000.1 | 12 - 13 | 1335 | 110 | |
XM_011526700.2 | XP_011525002.1 | 12 - 13 | 1281 | 110 | |
XM_011526701.2 | XP_011525003.1 | 7 - 8 | 884 | 110 | |
XM_017026550.1 | XP_016882039.1 | 11 - 12 | 1177 | 110 | |
GenBank mRNA | AB209773.1 | - | 9 - 10 | 1029 | 110 |
AF103939.1 | - | 12 - 13 | 1287 | 110 | |
AK026616.1 | - | 10 - 11 | 991 | 110 | |
AK225043.1 | - | 12 - 13 | 1279 | 110 | |
AK294668.1 | - | 15 - 16 | 1750 | 110 | |
AK295905.1 | - | 15 - 16 | 1832 | 110 | |
AK296258.1 | - | 15 - 16 | 1761 | 110 | |
AL096717.1 | - | 12 - 13 | 1264 | 110 | |
BC029304.1 | - | 15 - 16 | 1995 | 110 |