Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005727.3 | NP_005718.2 | 2 - 3 | 463 | 121 |
XM_011540460.2 | XP_011538762.1 | 2 - 3 | 437 | 121 | |
GenBank mRNA | AF065388.1 | - | 1 - 2 | 110 | 121 |
AF133425.1 | - | 1 - 2 | 112 | 121 | |
AK313774.1 | - | 2 - 3 | 453 | 121 | |
BC007290.1 | - | 1 - 2 | 116 | 121 | |
BC013404.2 | - | 1 - 2 | 114 | 121 | |
BM908049.1 | - | 2 - 3 | 466 | 121 |