Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288773.2 | NP_001275702.2 | 3 - 4 | 768 | 75 |
NM_001290113.1 | NP_001277042.1 | 2 - 3 | 518 | 75 | |
XM_011527450.1 | XP_011525752.1 | 3 - 4 | 806 | 75 | |
XM_011527452.1 | XP_011525754.1 | 3 - 4 | 768 | 75 | |
XM_017027434.1 | XP_016882923.1 | 2 - 3 | 733 | 75 | |
XM_017027435.1 | XP_016882924.1 | 2 - 3 | 733 | 75 | |
XM_017027436.1 | XP_016882925.1 | 6 - 7 | 1915 | 75 | |
XM_017027437.1 | XP_016882926.1 | 2 - 3 | 2948 | 75 | |
GenBank mRNA | AK023602.1 | - | 3 - 4 | 768 | 75 |
BC012001.1 | - | 2 - 3 | 518 | 75 |