Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030578.3 | NP_085055.2 | 1 - 2 | 222 | 106 |
GenBank mRNA | BC004157.2 | - | 1 - 2 | 181 | 106 |
BC004444.1 | - | 1 - 2 | 181 | 106 | |
BM833644.1 | - | 1 - 2 | 195 | 106 |