Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135585.1 | NP_001129057.1 | 3 - 4 | 592 | 73 |
NM_003039.2 | NP_003030.1 | 3 - 4 | 592 | 73 | |
XM_005263491.3 | XP_005263548.1 | 4 - 5 | 390 | 73 | |
XM_017002133.1 | XP_016857622.1 | 5 - 6 | 618 | 73 | |
XM_017002134.1 | XP_016857623.1 | 5 - 6 | 584 | 73 | |
XM_017002135.1 | XP_016857624.1 | 5 - 6 | 678 | 73 | |
XM_017002136.1 | XP_016857625.1 | 5 - 6 | 654 | 73 | |
XM_017002137.1 | XP_016857626.1 | 6 - 7 | 690 | 73 | |
XM_017002138.1 | XP_016857627.1 | 6 - 7 | 666 | 73 | |
XM_017002139.1 | XP_016857628.1 | 4 - 5 | 600 | 73 | |
XM_017002140.1 | XP_016857629.1 | 5 - 6 | 521 | 73 | |
XM_017002141.1 | XP_016857630.1 | 4 - 5 | 423 | 73 | |
GenBank mRNA | AF451839.1 | - | 1 - 2 | 105 | 73 |
AF479408.1 | - | 1 - 2 | 105 | 73 | |
AK289849.1 | - | 3 - 4 | 348 | 73 | |
AK290398.1 | - | 3 - 4 | 382 | 73 | |
AK294368.1 | - | 3 - 4 | 378 | 73 | |
AK302047.1 | - | 3 - 4 | 233 | 73 | |
AK302057.1 | - | 6 - 7 | 516 | 73 | |
BC001692.2 | - | 3 - 4 | 351 | 73 | |
BC001820.2 | - | 3 - 4 | 351 | 73 | |
BC035878.1 | - | 3 - 4 | 379 | 73 | |
M55531.1 | - | 3 - 4 | 375 | 73 |