Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018198.3 | NP_060668.2 | 2 - 3 | 309 | 102 |
GenBank mRNA | AF306695.1 | - | 2 - 3 | 223 | 102 |
AK298378.1 | - | 2 - 3 | 195 | 102 | |
AL109978.1 | - | 1 - 2 | 131 | 102 | |
AL833841.1 | - | 2 - 3 | 183 | 102 | |
AY336749.1 | - | 2 - 3 | 219 | 102 | |
BC006086.1 | - | 2 - 3 | 205 | 102 | |
BC008772.1 | - | 2 - 3 | 210 | 102 | |
BC014145.2 | - | 2 - 3 | 217 | 102 | |
DB461237.1 | - | 2 - 3 | 311 | 102 |