Assay Design



Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018198.3 NP_060668.2 7 - 8 802 62
XM_017001745.1 XP_016857234.1 6 - 7 655 62
GenBank mRNA AF306694.1 - 7 - 8 701 62
AF306695.1 - 7 - 8 716 62
AK001599.1 - 7 - 8 711 62
AK294539.1 - 6 - 7 625 62
AK298378.1 - 7 - 8 688 62
AL109978.1 - 6 - 7 624 62
AL833841.1 - 7 - 8 676 62
AY336749.1 - 7 - 8 712 62
BC006086.1 - 7 - 8 698 62
BC008772.1 - 7 - 8 703 62
BC014145.2 - 7 - 8 710 62

View Full Product Details